n-Hexane, PESTINORM® Supra Trace for MOSH/MOAH analysis

Supplier: VWR Chemicals
Danger

Synonyms: Hexyl hydride, Hexanen, Hexane

Filtered through a 0.2 µm filter, packaged under inert gas.

VWRC85957.320EA 373 SGD
VWRC85957.320
n-Hexane, PESTINORM® Supra Trace for MOSH/MOAH analysis
n-Hexane

Mineral oils are almost universally present in our environment. The main compounds of interest are mineral oil saturated hydrocarbons (MOSH) and mineral oil aromatic hydrocarbons (MOAH).
Both types are easily absorbed by the human body from food and can accumulate in body fat and in some organs.


  • Specifically designed and tested for the MOSH/MOAH analysis
  • Tested and guaranteed by HPLC/UV – GC-FID and GC-MS to have less than 3 ng/ml, and by GC-ECD less than 3 pg/ml of any peak in the full range from C10 to C50 of the MOSH/MOAH impurities

Formula: H₃C(CH₂)₄CH₃
MW: 86.18 g/mol
Boiling Pt: 69 °C (1013 hPa)
Melting Pt: –94,3 °C
Density: 0.659 g/cm³ (20 °C)
Flash Pt: –22 °C
Storage Temperature: Ambient
MDL Number: MFCD00009520
CAS Number: 110-54-3
EINECS: 203-777-6
UN: 1208
ADR: 3 II
Merck Index: 13,04712
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fabd698969eb09b591de207cbcb599c33689bf7f&vid=c117cbd0b3ff2cab0e7f8e5a4bf7108f938081d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear colorless liquid
Identity (IR) Complies
Color (APHA, Pt-Co) max. 10
Assay (GC, on anhydrous basis) min. 99.0 %
Hexane isomers + methylcyclopentane min. 99.8 %
MOSH Test by GC/FID max. 5 ng/ml
MOAH Test by LC/UV max. 5 ng/ml
GC/ECD 1,2,4-TCB to Deca-PCB (as Lindane) max. 3 pg/ml
Water (KF) max. 0.02 % w/w
Residue after evaporation max. 0.0003 % w/w


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR