Standard solution set (potassium dichromate solutions) for calibration of UV/Visible spectrophotometers
Supplier: VWR Chemicals
VWRC85969.001EA 815 SGD
VWRC85969.001
Standard solution set (potassium dichromate solutions) for calibration of UV/Visible spectrophotometers
Standards Colour Reference and Spectroscopy Standards
A set for linearity check of the absorbance of spectrophotometers.
A set of reagents for calibration of spectrophotometers, to provide the user with a range two different concentrations of potassium dichromate and additional sulphuric acid as a reference solution.
Set contains:
1×250 ml potassium dichromate 50 mg/L in H₂SO₄ (0,005 M)
1×250 ml potassium dichromate 100 mg/L in H₂SO₄ (0,005 M)
2×250 ml sulphuric acid (0,005 M)