Carbon disulphide ≥99.90%, SPECTRONORM® for spectroscopy

Supplier: VWR Chemicals
Danger


Packaged under nitrogen.

VWRC85971.290EA 600 SGD
VWRC85971.290
Carbon disulphide ≥99.90%, SPECTRONORM® for spectroscopy
Carbon disulphide

SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.

Formula: CS₂
Boiling Pt: 46,2 °C (1013 hPa)
Melting Pt: –111,6 °C
Density: 1,2632 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00011321
CAS Number: 75-15-0
EINECS: 200-843-6
UN: 1131
ADR: 3 (6.1),I
Merck Index: 13,01821
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7fe0c906449b3b5461b1bac971c583c92c0b41c&vid=59e9028fbb4d1c2acb6f98101726e8ec520e0aa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay min. 99.90%
Water 0.01%
Evaporation residue max. 0.0010%
Benzene (GC) max. 0.0030%
Toluene (GC) max. 0.0010%
Xylene (GC) max. 0.0010%
Alkalinity (meq/g) max. 0.0002
Acidity (meq/g) max. 0.0005
Color (APHA) max. 10
UV Transmission (382 nm) min. 10.00%
UV Transmission (388 nm) min. 35.00%
UV Transmission (403 nm) min. 80.00%
UV Transmission (410 nm) min. 90.00%
UV Transmission (500 nm) min. 99.00%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR