N,N-Diethyl-p-phenylenediammonium sulphate 99, AnalaR NORMAPUR® zA

Supplier: VWR Chemicals
Warning

Synonyms: N,N-Diethyl-p-phenylenediamine sulphate, N,N-Diethyl-1,4-phenylenediammonium sulphate, 4-Amino-N,N-diethylaniline sulphate, N1,N1-Diethylbenzene-1,4-diamine sulphate, N,N-Diethylbenzene-1,4-diammonium sulphate

VWRC87026.180EA 675 SGD
VWRC87026.180 VWRC87026.460
N,N-Diethyl-p-phenylenediammonium sulphate 99, AnalaR NORMAPUR® zA
N,N-Diethyl-p-phenylenediammonium sulphate
Formula: C₁₀H₁₈N₂O₄S
MW: 262.33 g/mol
Boiling Pt: 274…275 °C (1013 hPa)
Melting Pt: 184…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00012993
CAS Number: 6283-63-2
EINECS: 228-500-6

Order Now

Specification Test Results

Assay Min. 99 %
Heavy metals (as Pb) Max. 10 ppm
Ignition residue (SO4) Max. 0,05 %
Water Max. 0,5 %
Fe (Iron) Max. 10 ppm

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR