VWR® ARISTAR®, Osmium Standard Solution for ICP
Supplier: VWR Chemicals
VWRC87051.180EA 944 SGD
VWRC87051.180
VWR® ARISTAR®, Osmium Standard Solution for ICP
Standards Cationic Standards Osmium Standards
  • Produced from high purity OsCl₃
  • Certified reference material
  • Traceable to NIST
  • Certified Value and uncertainty 1000,0 ± 0,5 mg/L
Density 1,060 g/cm³ at 20 °C.
Caution: Storage at ambient temperatures.