Methyl red solution_SC Reag. Ph. Eur. 1055101

Supplier: VWR Chemicals

Synonyms: 2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid

VWRC87703.180EA 124 SGD
VWRC87703.180
Methyl red solution_SC Reag. Ph. Eur. 1055101
Methyl red
Formula: C₁₅H₁₅N₃O₂
MW: 269.3 g/mol
MDL Number: MFCD00002425
CAS Number: 493-52-7
Merck Index: 12,06199

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR