Phenolphthalein 0,1% in ethanol 80% Reag. Ph. Eur. 1063702

Supplier: VWR Chemicals

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

VWRC87705.180EA 118 SGD
VWRC87705.180
Phenolphthalein 0,1% in ethanol 80% Reag. Ph. Eur. 1063702
Phenolphthalein

Ordering information: Normally manufactured to order. Please check with customer services.

Formula: C₂₀H₁₄O₄
MDL Number: MFCD00005913
CAS Number: 77-09-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR