Alizarin red S (sodium salt) solution Reag. Ph. Eur. 1002601

Supplier: VWR Chemicals

VWRC87763.180EA 191 SGD
VWRC87763.180
Alizarin red S (sodium salt) solution Reag. Ph. Eur. 1002601
Alizarin Red S (sodium salt)

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₁₄H₇NaO₇S
MDL Number: MFCD00013049
CAS Number: 130-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR