Ammonium carbonate 1 mol/l in aqueous solution Reag. Ph. Eur. 1005201

Supplier: VWR Chemicals

Synonyms: Salt of hartshorn

VWRC87768.290EA 606 SGD
VWRC87768.290
Ammonium carbonate 1 mol/l in aqueous solution Reag. Ph. Eur. 1005201
Ammonium carbonate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: (NH₄)₂CO₃
MW: 96,09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR