Ammonium chloride 2 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1005301

Supplier: VWR Chemicals

Synonyms: Ammonium muriate, Salmiac

VWRC87769.290EA 301 SGD
VWRC87769.290
Ammonium chloride 2 mol/l (2 N) in aqueous solution Reag. Ph. Eur. 1005301
Ammonium chloride

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: NH₄Cl
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR