Bromocresol purple solution Reag. Ph. Eur. 1012701

Supplier: VWR Chemicals

Synonyms: 5',5''-Dibromo-o-cresolsulphonphthalein, 3,3-Bis(3-bromo-4-hydroxy-5-methylphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

VWRC87793.180EA 111 SGD
VWRC87793.180
Bromocresol purple solution Reag. Ph. Eur. 1012701
Bromocresol purple
Formula: C₂₁H₁₆Br₂O₅S
MW: 540.23 g/mol
Melting Pt: 242
MDL Number: MFCD00011681
CAS Number: 115-40-2
Merck Index: 13,01371

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR