Copper(II) sulphate 0.5 mol/l Reag. Ph. Eur. 1022501

Supplier: VWR Chemicals
Warning

VWRC87808.290EA 347 SGD
VWRC87808.290
Copper(II) sulphate 0.5 mol/l Reag. Ph. Eur. 1022501
Copper (II) sulphate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: CuSO₄
Storage Temperature: Ambient
MDL Number: MFCD00010981
CAS Number: 7758-98-7
EINECS: 231-847-6
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR