Ammonium iron(II) sulphate 100 g/l solution R2 Reag. Ph. Eur. 1037702

Supplier: VWR Chemicals

Synonyms: di-Ammonium iron (II) sulphate, Ammonium ferrous sulphate

VWRC87830.290EA 277 SGD
VWRC87830.290
Ammonium iron(II) sulphate 100 g/l solution R2 Reag. Ph. Eur. 1037702
Ammonium iron (II) sulphate

Other name: Ferric ammonium sulphate solution R2


Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: H₈FeN₂O₈S₂
MW: 284.05 g/mol
Melting Pt: 100
Density: 1.86
Storage Temperature: Ambient
MDL Number: MFCD00051039
CAS Number: 10045-89-3
UN: 3264
ADR: 8 II
Merck Index: 12,00552

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR