Iron(III) chloride 13 g/l in aqueous solution Reag. Ph. Eur. 1037802

Supplier: VWR Chemicals
Warning

Synonyms: Iron trichloride, Ferric chloride

VWRC87834.290EA 237 SGD
VWRC87834.290
Iron(III) chloride 13 g/l in aqueous solution Reag. Ph. Eur. 1037802
Iron (III) chloride

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: FeCl₃
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR