Iron(III) chloride solution R3 Reag. Ph. Eur. 1037803

Supplier: VWR Chemicals
Danger

VWRC87835.180EA 137 SGD
VWRC87835.180
Iron(III) chloride solution R3 Reag. Ph. Eur. 1037803
Iron (III) chloride

Ferric chloride solution R3 Reag. Ph. Eur. 1037803


Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: FeCl₃
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR