Hydroxylamine solution, alcoholic Reag. Ph. Eur. 1044301

Supplier: VWR Chemicals

VWRC87847.180EA 326 SGD
VWRC87847.180
Hydroxylamine solution, alcoholic Reag. Ph. Eur. 1044301
Hydroxylamine

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: H₃NO
MDL Number: MFCD00044522
CAS Number: 7803-49-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR