Oxalic acid 0,5% sulphuric acid_50 Reag. Ph. Eur. 1061401

Supplier: VWR Chemicals
Danger

Synonyms: Ethanedioic acid

VWRC87883.290EA 370 SGD
VWRC87883.290
Oxalic acid 0,5% sulphuric acid_50 Reag. Ph. Eur. 1061401
Oxalic acid

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: C₂H₂O₄
MW: 90.04 g/mol
Boiling Pt: 365
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 2796
ADR: 8 II
Merck Index: 13,06980

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR