Phosphotungstic acid 22% in orthophosphoric acid 11% Reag. Ph. Eur. 1065200

Supplier: VWR Chemicals
Danger

VWRC87896.180EA 210 SGD
VWRC87896.180
Phosphotungstic acid 22% in orthophosphoric acid 11% Reag. Ph. Eur. 1065200
Phosphotungstic acid

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: H₃O₄₀PW₁₂
CAS Number: 1343-93-7
EINECS: 215-682-7
UN: 3264
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR