Potassium chloride 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1069101

Supplier: VWR Chemicals

VWRC87898.290EA 224 SGD
VWRC87898.290
Potassium chloride 0.1 mol/l (0.1 N) in aqueous solution Reag. Ph. Eur. 1069101
Potassium chloride

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: KCl
Storage Temperature: Ambient
MDL Number: MFCD00011360
CAS Number: 7447-40-7
EINECS: 231-211-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR