Potassium heptaiodobismuthate 0,5% acetic acid_diluted with barium chloride Reag. Ph. Eur. 1070605

Supplier: VWR Chemicals

VWRC87916.180EA 628 SGD
VWRC87916.180
Potassium heptaiodobismuthate 0,5% acetic acid_diluted with barium chloride Reag. Ph. Eur. 1070605
Potassium heptaiodobismuthate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: BiI₇K₄
MW: 1253.71 g/mol
CAS Number: 41944-01-8
EINECS: 255-595-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR