Potassium permanganate 30 g/l ortho-phosphoric acid_22 Reag. Ph. Eur. 1070901

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

VWRC87918.180EA 106 SGD
VWRC87918.180
Potassium permanganate 30 g/l ortho-phosphoric acid_22 Reag. Ph. Eur. 1070901
Potassium permanganate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 1805
ADR: 8 III
Merck Index: 12,07824

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR