Potassium permanganate 3% in aqueous solution Reag. Ph. Eur. 1070902

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

VWRC87919.290EA 236 SGD
VWRC87919.290
Potassium permanganate 3% in aqueous solution Reag. Ph. Eur. 1070902
Potassium permanganate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: KMnO₄
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
EINECS: 231-760-3
UN: 3082
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR