Sodium carbonate 4% in sodium hydroxide solution 0.2 mol/l Reag. Ph. Eur. 1079303

Supplier: VWR Chemicals

Synonyms: Calcined soda, Carbonic acid disodium salt

VWRC87930.290EA 350 SGD
VWRC87930.290
Sodium carbonate 4% in sodium hydroxide solution 0.2 mol/l Reag. Ph. Eur. 1079303
Sodium carbonate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR