Sodium hydrogen carbonate 0.5 mol/l in aqueous solution Reag. Ph. Eur. 1081301

Supplier: VWR Chemicals

VWRC87932.290EA 268 SGD
VWRC87932.290
Sodium hydrogen carbonate 0.5 mol/l in aqueous solution Reag. Ph. Eur. 1081301
Sodium hydrogen carbonate

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: NaHCO₃
Storage Temperature: Ambient
MDL Number: MFCD00003528
CAS Number: 144-55-8
EINECS: 205-633-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6b1b80d2afb3deab0c4ca836a921fcaad637890&vid=1163b37d9cf1c10fe0ed252e02a51fe12d6b50dd&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR