Water Reag. Ph. Eur. 1095503 for chromatography

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

VWRC87960.290EA 173 SGD
VWRC87960.290
Water Reag. Ph. Eur. 1095503 for chromatography
Water

Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR