VWR® Fixing Solution
Supplier: VWR Chemicals
VWRC87970.260EA 188 SGD
VWRC87970.260
VWR® Fixing Solution
Stains Microscopy and staining reagents
Fixing solution with 0,02% formaldehyde in methanol 50% (v/v)).
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.