Water, PESTINORM® for headspace gas chromatography

Supplier: VWR Chemicals

VWRC88251.290EA 209 SGD
VWRC88251.290
Water, PESTINORM® for headspace gas chromatography
Water
Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Clear colorless liquid
Color (APHA, Pt-Co) max. 5
Acidity (as Acetic acid) max. 0.0005 %
Alkalinity (as Ammonia) max. 0.0001 %
GC-HS (Each class 1 solvent) max. 1 ug/g
GC-HS (Each class 2 solvent) max. 10 ug/g
GC-HS (Each class 3 solvent) max. 50 ug/g
Headspace test for O.V.I's Complying
Suitable as solvent for the analysis of residual solvents of ICH classes 1, 2 and 3
according to Ph Eur and USP.
Formic acid, acetic acid, N,N-dimethylacetamide, N,N-dimethylformamide and dimethyl
sulfoxide are not detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR