Hydrochloric acid 37% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytical reagent

Supplier: VWR Chemicals
Danger

VWRC88372.320EA 80.4 SGD
VWRC88372.320
Hydrochloric acid 37% (max. 0.005 ppm Hg), AnalaR NORMAPUR® analytical reagent
Hydrochloric acid
Formula: HCl
MDL Number: MFCD00011324
CAS Number: 7647-01-0
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Assay 37 - 38%
Ammonium (NH4) max. 3 ppm
Free Chlorine max. 0,5 ppm
Mercury (Hg) max. 5 ppb
Residue after ignition max. 5 ppm
Sulphate (SO4) max. 1 ppm
Sulfite (SO3) max. 2 ppm
Aluminium (Al) max. 0,05 ppm
Arsenic (As) max. 0,05 ppm
Barium (Ba) max. 0,02 ppm
Beryllium (Be) max. 0,01 ppm
Cadmium (Cd) max. 0,02 ppm
Calcium (Ca) max. 0,5 ppm
Chromium (Cr) max. 0,05 ppm
Cobalt (Co) max. 0,01 ppm
Copper (Cu) max. 0,01 ppm
Germanium (Ge) max. 0,1 ppm
Iron (Fe) max. 0,2 ppm
Lead (Pb) max. 0,05 ppm
Lithium (Li) max. 0,01 ppm
Magnesium (Mg) max. 0,05 ppm
Manganese (Mn) max. 0,01 ppm
Molybdenum (Mo) max. 0,02 ppm
Nickel (Ni) max. 0,05 ppm
Potassium (K) max. 0,1 ppm
Silver (Ag) max. 0,01 ppm
Sodium (Na) max. 0,3 ppm
Strontium (Sr) max. 0,01 ppm
Thallium (Tl) max. 0,05 ppm
Titanium (Ti) max. 0,05 ppm
Vanadium (V) max. 0,01 ppm
Zinc (Zn) max. 0,5 ppm
Zirconium (Zr) max. 0,05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR