VWR®, Coulometry NewGen Reagent AG-FI for coulometric Karl Fischer titration
Supplier: VWR Chemicals
VWRC88425.260EA 474 SGD
VWRC88425.260
VWR®, Coulometry NewGen Reagent AG-FI for coulometric Karl Fischer titration
Karl Fischer Titrants and Reagents
Anolyte for coulometric Karl Fischer titration, for cells with and without diaphragm, Methanol-based, free of Imidazole (free of CMR substances).
  • Free of imidazole
  • Free of CMR substances
Specification:
Suitability (for KF-Titration): complying
Water content: 5-50 ppm