VWR®, Karl Fischer NewGen Solvent-FI for volumetric two-component KF titration
Supplier: VWR Chemicals
VWRC88426.290EA 415 SGD
VWRC88426.290
VWR®, Karl Fischer NewGen Solvent-FI for volumetric two-component KF titration
Karl Fischer Titrants and Reagents
Medium for volumetric two-component Karl Fischer titration (methanol-based), Free of imidazole (free of CMR substances)
  • free of imidazole
  • free of CMR substances
Specification:
Suitability (for KF-Titration): complying
Water content: 0.010 - 0.020% (v/v)