Sodium nitrite 0.1 N, AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Diazotizing salts

VWRC89295.320EA 125 SGD
VWRC89295.320
Sodium nitrite 0.1 N, AVS TITRINORM volumetric solution
Sodium nitrite
Formula: NaNO₂
MDL Number: MFCD00011118
CAS Number: 7632-00-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR