Potassium chloride 0.2 mol/l (0.2 N) in aqueous solution volumetric solution

Supplier: VWR Chemicals

Synonyms: Potassiumchloride

VWRC89511.290EA 80.3 SGD
VWRC89511.290
Potassium chloride 0.2 mol/l (0.2 N) in aqueous solution volumetric solution
Potassium chloride
Formula: KCl
MDL Number: MFCD00011360
CAS Number: 7447-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR