Oxalic acid 0.005 mol/l (0.01 N), AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

VWRC89619.290EA 71.9 SGD
VWRC89619.290
Oxalic acid 0.005 mol/l (0.01 N), AVS TITRINORM volumetric solution
Oxalic acid
Formula: C₂H₂O₄
MDL Number: MFCD00002573
CAS Number: 144-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR