VWR® PESTINORM®, LC Multiresidue Pesticide Standard (Mix 10) - 1 component, CRM
Supplier: VWR Chemicals
VWRC91132.001EA 112 SGD
VWRC91132.001
VWR® PESTINORM®, LC Multiresidue Pesticide Standard (Mix 10) - 1 component, CRM
Standards Organic Standards
A standard, typically used in food testing, Carbendazim [CAS:10605-21-7] 100 ug/ml in Methanol
All mixes are supplied with detailed certificate of analysis, chromatogram and 100% shelf-life. The CoAs provide full traceability of preparation, including the dissolution of the raw materials (with their purity), the preparation of the intermediate solutions and the preparation of the final solutions.
Certifications: Certified Reference Material (CRM) acc. ISO 17025 / ISO 17034
Caution: Storage temperature: Freezer (–20 °C)