VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Supplier: VWR Chemicals
VWRC91027.050EA 310 SGD
VWRC91027.050 VWRC91028.050 VWRC91031.050 VWRC91031.010 VWRC91027.010 VWRC91072.050 VWRC91073.050 VWRC91074.010 VWRC91074.050 VWRC91087.002 VWRC91087.010 VWRC91087.050 VWRC91088.001 VWRC91088.010 VWRC91088.050 VWRC91090.001 VWRC91090.010 VWRC91090.050 VWRC91091.001 VWRC91091.002 VWRC91091.010 VWRC91072.010 VWRC91250.001
VWR® PESTINORM®, Fatty Acid Methyl Esters (FAME) Standards for GC
Standards Chromatography Standards
Avantor offers an extensive range of FAME standards as Certified Reference Materials under the well-known PESTINORM® brand.
  • All products are CRM acc ISO/IEC 17025 and ISO 17034
  • All products are NEAT
  • 24 month shelf life
Esterification of fatty acids is an important tool to determine content and characterise fats and oils in food, especially for measurement by GC methods. The simple derivatisation to create the methyl ester analogue is also used widely as a preliminary technique for assessing the quality and purity of biofuels. VWR® offers a comprehensive range of fatty acid methyl esters (FAME) Certified Reference Materials for calibration of GC instruments, for validation of analytical methods, preparation of 'working reference samples' or for detection limit and linearity studies.
Certifications: ISO 17025 / 17034
Order Now
SPECIFICATIONS
CAS No. 111-11-5 112-61-8 124-10-7 106-70-7 106-73-0 1731-94-8 1120-28-1 929-77-1 2442-49-1 624-24-8 301-00-8
Environmentally Preferable