Molybdic acid 85 zS

Supplier: VWR Chemicals

VWRC91302.180EA 128 SGD
VWRC91302.180
Molybdic acid 85 zS
Molybdic acid
Formula: H₂MoO₄
MW: 161.95 g/mol
Boiling Pt: ∼300 °C (1013 hPa)
Melting Pt: 300 °C
Density: 3.1 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00036287
CAS Number: 7782-91-4
EINECS: 231-970-5
Merck Index: 13,06262

Order Now

Specification Test Results

Assay Min 85%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR