Potassium permanganate concentrate for dilution to 0.02 mol (0.1N) aqueous solution, ConvoL NORMADOSE® Volumetric

Supplier: VWR Chemicals
Danger

Synonyms: Permanganic acid potassium salt

conc_1l

VWRC91352.600EA 347 SGD
VWRC91352.600
Potassium permanganate concentrate for dilution to 0.02 mol (0.1N) aqueous solution, ConvoL NORMADOSE® Volumetric
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
Merck Index: 12,07824

Order Now

Specification Test Results

Molarity 0.0199 - 0.0201 mol/l (20°C)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR