Potassium permanganate concentrate for dilution to 0.002 mol (0.01N) aqueous solution, ConvoL NORMADOSE® Volumetric

Supplier: VWR Chemicals

Synonyms: Permanganic acid potassium salt

conc_1l

VWRC91353.600EA 194 SGD
VWRC91353.600
Potassium permanganate concentrate for dilution to 0.002 mol (0.01N) aqueous solution, ConvoL NORMADOSE® Volumetric
Potassium permanganate
Formula: KMnO₄
MW: 158.03 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011364
CAS Number: 7722-64-7
Merck Index: 12,07824

Order Now

Specification Test Results

Molarity 0.00199 - 0.00201 mol/l (20°C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR