VWR®, Perfluorohexane-1-sulphonic acid (PFHxS), 2 µg/ml in MeOH, Certified Reference Material (CRM)
Supplier: VWR Chemicals
VWRC91407.010EA 423 SGD
VWRC91407.010
VWR®, Perfluorohexane-1-sulphonic acid (PFHxS), 2 µg/ml in MeOH, Certified Reference Material (CRM)
Standards Organic Standards
Per- and polyfluoroalkyl substances (PFAS) are amongst the primary emerging contaminants of concern. The detection and quantification of known PFAS and the discovery of unknown PFAS substances has never been more important.
VWR® is supplying a comprehensive range of standards and CRMs to enable this environmental analysis.
Certifications: Certified Reference Material (CRM) ISO 17025 / ISO 17034