Water 99,9, HiPerSolv CHROMANORM®, PFAS grade LC-MS

Supplier: VWR Chemicals

Synonyms: Hydrogen oxide

Filter 0,2 um_Nitrogen_filled

VWRC92499.320EA 681 SGD
VWRC92499.320
Water 99,9, HiPerSolv CHROMANORM®, PFAS grade LC-MS
Water

VWR® PFAS grade LC-MS solvents undergo a rigorous use test to ensure suitability for analysis of poly- and perfluoroalkyl substances resulting in an ultra low background of PFAS analytes. Using complementary methods a broad range of PFAS analytes are quantified to their detection limits by LC-MS.


  • Specifically tested for PFAS analysis
  • Ultra low background solvent for LC-MS analysis of PFAS (poly- and perfluoroalkyl substances)

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Resistivity (at filling; @ 25°C) min. 18 Mohm
TOC (at filling) max. 10 ppb
pH (at filling; @25°C) 5.5 - 8.0
Evaporation residue max. 0.00005%
Gradient use test (205 nm) max. 0.002 AU
Gradient use test (254 nm) max. 0.0005 AU
Baseline drift (205 nm) max. 0.02 AU
Ag (Silver) max. 10 ppb
Al (Aluminium) max. 25 ppb
Ca (Calcium) max. 25 ppb
Cu (Copper) max. 10 ppb
Fe (Iron) max. 10 ppb
K (Potassium) max. 10 ppb
Mg (Magnesium) max. 10 ppb
Mn (Manganese) max. 10 ppb
Na (Sodium) max. 50 ppb
Ni (Nickel) max. 10 ppb
Pb (Lead) max. 10 ppb
Zn (Zinc) max. 10 ppb
Suitability for PFAS analysis passes test
Suitability for LC-MS passes test
manufactured and filled under nitrogen
Filtered to 0.2 µm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR