Selenium reagent mixture for the determination of nitrogen acc. to Wieninger
Supplier: VWR Chemicals
Warning
VWRC92504.290EA 225 SGD
VWRC92504.290
Selenium reagent mixture for the determination of nitrogen acc. to Wieninger
Selenium reagent mixture
Order Now
Specification Test Results
Na2SO4 (sodium sulfate) 97.00%
CuSO4 (copper sulfate) 1.50%
Se (selenium) 1.50%
Application test (total nitrogen) conforms