VWR®, Glass Beads
Supplier: VWR Chemicals
VWRC92509.260EA 56.7 SGD
VWRC92509.260 VWRC92505.260 VWRC92506.260 VWRC92507.260 VWRC92508.260
VWR®, Glass Beads
Beads Glass Beads
Soda-lime glass, solid.
  • Material: Soda-lime glass
  • Suitability: Not suitable for heat applications
  • Sterility: Non sterile
Can be used for many different applications in the laboratory. In microbiology laboratories they are used for the production of single cell suspensions or for plating. In chemical laboratories they are used as filling material for distillation columns. They can also be used to clean round bottom vessels.
Physical & Chemical Characteristics
Specific weight: 2.500 ± 40 kg/m3
Coefficient of thermal extension: (9.2 ± 0.4) . 10-6 K-1More Product Information