VWR® Oxidase Swabs in a Box
Supplier: VWR Chemicals
VWRC942152ZFEA 78.4 SGD
VWRC942152ZF
VWR® Oxidase Swabs in a Box
Microbiology Test Systems Enzymatic Tests
Swabs imprenated with reagent for the detection and verification of bacterial cytochrome oxidase.
  • 2×25 swabs
Caution: *The product(s) below are outside the scope of the EU Directive 2019/904.
Order Now
SPECIFICATIONS
Shelf life 360 days
Environmentally Preferable
Storage temperature 2...14 °C