Alizarin red S (sodium salt)

Supplier: VWR Chemicals
Warning

VWRC9436-25GEA 161 SGD
VWRC9436-25G
Alizarin red S (sodium salt)
Alizarin Red S (sodium salt)
Formula: C₁₄H₇NaO₇S
Storage Temperature: Ambient
MDL Number: MFCD00013049
CAS Number: 130-22-3
EINECS: 204-981-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR