Orthoboric acid 40 g/l in aqueous solution with indicator, VOLUSOL®

Supplier: VWR Chemicals

Synonyms: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

VWRC95097.5000EA 56.5 SGD
VWRC95097.5000
Orthoboric acid 40 g/l in aqueous solution with indicator, VOLUSOL®
ortho-Boric acid
Formula: B(OH)₃
Storage Temperature: Ambient
MDL Number: MFCD00011337
CAS Number: 10043-35-3
EINECS: 233-139-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (W/V) 3.9 - 4.2 %
Appearance Clear red liquid
Sensitivity Passes test
Density (20/4) 1.000 - 1.020


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR