Hydrochloric acid 0.02 mol/l (0.02 N), AVS TITRINORM volumetric solution

Supplier: VWR Chemicals

Synonyms: Hydrochloric acid solution, Muriatic acid

VWRC98052.296EA 16.2 SGD
VWRC98052.296
Hydrochloric acid 0.02 mol/l (0.02 N), AVS TITRINORM volumetric solution
Hydrochloric acid
Formula: HCl
MW: 36.46
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 III

Order Now

Specification Test Results

Titer (20°C; real value 0.2 % accuracy) 0.01996 - 0.02004 mol/l
NIST traceable Confirmed


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR