Ethanol 70% ≥70% (v/v), GPR RECTAPUR®

Supplier: VWR Chemicals
Danger

Synonyms: Ethyl alcohol

VWRC98193.418EA 456 SGD
VWRC98193.418 VWRC98193.418DP
Ethanol 70% ≥70% (v/v), GPR RECTAPUR®
Ethanol

Caution: WARNING : This alcohol is regulated. Please contact VWR. For duty paid orders in the UK please add DP to the product code.

Formula: C₂H₅OH
Density: 0,85 g/cm³ (20 °C)
Flash Pt: 22 °C
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Assay (on anhydrous substance) Min. 99 %
Assay (V/V) 69.0-72.0 %
Evaporation residue Max. 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR