Sodium chloride regeneration salt, TECHNICAL for water softening

Supplier: VWR Chemicals

Synonyms: Saline, NaCl, Sea salt, Halite

VWRC99062.4000EA 36.1 SGD
VWRC99062.4000
Sodium chloride regeneration salt, TECHNICAL for water softening
Sodium chloride
Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413 °C (1013 hPa)
Melting Pt: 801
Density: 2.16 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003477
CAS Number: 7647-14-5
EINECS: 231-598-3
Merck Index: 14,08599

Order Now

Specification Test Results

Identification Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR