VWR®, Flash point reference standard, Abel Closed Cup, jet aviation fuel
Supplier: VWR Chemicals
VWRC99116.230EA 1280 SGD
VWRC99116.230
VWR®, Flash point reference standard, Abel Closed Cup, jet aviation fuel
Standards Petroleum Industry Standards
Flash point Certified Reference Material for the calibration and verification of laboratory instruments used in testing petroleum and derivative products for Abel Closed Cup (IP 170, ISO 13736).
  • Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
  • Fully traceable to international standards
  • Supplied with MSDS
  • Tamper-evident security packaging
  • 12 month shelf life
Certifications: UKAS ISO 17025 and ISO 17034.
Caution: The flash point given in the ordering table below is the nominal flash point - actual batch values are given in the certification documents supplied with the product and also on the label.