VWR®, Pre-weighed dehydrated culture media in sachets
Supplier: VWR Chemicals
VWRCS246SAC5EA 54.3 SGD
VWRCS246SAC5 VWRCA887SAC5 VWRCS378SAC5 VWRCB202SAC5 VWRCB455SAC5 VWRCA990SAC5
VWR®, Pre-weighed dehydrated culture media in sachets
Media for Microbiology Dehydrated Media for Microbiology
Dehydrated culture media in small sachets ready to make up exactly 500 ml of medium.
  • Quick: Simply open the sachet and you have the right quantity of powder to prepare 500 ml of medium
  • Safe: No more need to transfer and weigh powder
  • Always fresh media for best quality: All the dehydrated media inside the sachet is always used immediately after opening the packaging
  • Space saving: You can have tens of dehydrated media stored in a very small space inside your laboratory
  • Saving money: You buy only the quantity of dehydrated media that you really need
Every pack contains 5 sachets each of which allows to prepare 500 ml of medium.
Ordering information: About 140 different culture media available.